Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317791.1 | NP_001304720.1 | 2 | 155 | ||
NM_144721.5 | NP_653322.1 | 2 | 155 | |||
XM_005262774.3 | XP_005262831.1 | 2 | 157 | |||
XM_006714109.3 | XP_006714172.1 | 2 | 157 | |||
XM_011531666.2 | XP_011529968.1 | 2 | 592 | |||
XM_011531667.2 | XP_011529969.1 | 2 | 115 | |||
XM_017007800.1 | XP_016863289.1 | 2 | 321 | |||
XM_017007801.1 | XP_016863290.1 | 2 | 157 | |||
GenBank mRNA | AK303658.1 | - | 2 | 115 | ||
AK313765.1 | - | 2 | 539 | |||
AL833350.1 | - | 2 | 115 | |||
BC022989.2 | - | 1 | 69 | 5113206 | ||
BX647579.1 | - | 2 | 115 | |||
DC308535.1 | - | 2 | 156 |