Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198150.2 | NP_937793.1 | 5 | 921 | ||
XM_005271904.3 | XP_005271961.1 | 6 | 985 | |||
GenBank mRNA | AK303855.1 | - | 5 | 973 | ||
AK316253.1 | - | 5 | 940 | |||
AL832711.1 | - | 4 | 604 | |||
AY358596.1 | - | 5 | 881 | |||
AY875939.1 | - | 5 | 763 | |||
BC036047.1 | - | 5 | 783 | |||
BC050392.1 | - | 6 | 931 | |||
BC130329.1 | - | 5 | 859 | 40146360 | ||
BC144161.1 | - | 5 | 859 |