Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000397.3 | NP_000388.2 | 5 | 458 | ||
GenBank mRNA | AK289753.1 | - | 5 | 443 | ||
AK301491.1 | - | 4 | 320 | |||
AK312929.1 | - | 5 | 418 | |||
BC032720.1 | - | 5 | 444 | 5505170 | ||
DA995835.1 | - | 5 | 458 | |||
KU178009.1 | - | 5 | 397 | |||
KU178010.1 | - | 5 | 397 | |||
X04011.1 | - | 5 | 412 |