Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC2HC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013623.2 | NP_001013645.1 | 5 | 475 | ||
GenBank mRNA | BC146950.1 | - | 5 | 475 | 9056878 | |
BC146969.1 | - | 5 | 475 | 9056897 | ||
BC171862.1 | - | 6 | 558 | 9054516 |