Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173515.2 | NP_775786.2 | 1, 2 | 568 | ||
XM_011535485.2 | XP_011533787.1 | 1, 2 | 3051 | |||
GenBank mRNA | AK055911.1 | - | 1, 2 | 608 | ||
AK309680.1 | - | 1, 2 | 174 | |||
AY328891.1 | - | 1, 2 | 568 | |||
AY328892.1 | - | 1, 2 | 568 | |||
AY328893.1 | - | 1, 2 | 568 | |||
BC060761.1 | - | 1, 2 | 490 | 5299236 |