Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017373.3 | NP_001017373.2 | 7 | 919 | ||
NM_001258275.2 | NP_001245204.1 | 9 | 1200 | |||
NM_001277185.1 | NP_001264114.1 | 6 | 701 | |||
XM_017010305.1 | XP_016865794.1 | 6 | 746 | |||
XM_017010306.1 | XP_016865795.1 | 5 | 959 | |||
XM_017010307.1 | XP_016865796.1 | 5 | 956 | |||
XM_017010308.1 | XP_016865797.1 | 5 | 612 | |||
XM_017010309.1 | XP_016865798.1 | 5 | 612 | |||
XM_017010310.1 | XP_016865799.1 | 5 | 612 | |||
GenBank mRNA | AK091351.1 | - | 9 | 1202 | ||
AK091882.1 | - | 7 | 919 | |||
AK302228.1 | - | 6 | 703 | |||
AK310075.1 | - | 5 | 955 | |||
BC029851.1 | - | 5 | 608 | |||
BC119757.1 | - | 5 | 608 | |||
BC119758.1 | - | 5 | 608 | |||
BC127653.1 | - | 5 | 606 | |||
BC127654.1 | - | 5 | 606 |