Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VKORC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284342.1 | NP_001271271.1 | 2 | 385 | ||
NM_173517.4 | NP_775788.2 | 3 | 495 | |||
XM_011515830.2 | XP_011514132.1 | 4 | 2291 | |||
XM_011515831.2 | XP_011514133.1 | 4 | 321 | |||
GenBank mRNA | AK124688.1 | - | 2 | 148 | ||
AK289522.1 | - | 3 | 408 | |||
AK304077.1 | - | 2 | 286 | |||
AY423045.1 | - | 3 | 414 | |||
BC027734.1 | - | 3 | 495 | 5093832 | ||
BX647631.1 | - | 3 | 428 | |||
CR627471.1 | - | 2 | 183 |