Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000771.3 | NP_000762.2 | 8 | 1188 | ||
XM_017015758.1 | XP_016871247.1 | 9 | 1608 | |||
GenBank mRNA | AK289420.1 | - | 8 | 1188 | ||
AK298458.1 | - | 8 | 1188 | |||
BC125054.1 | - | 8 | 1163 | 40116637 | ||
D00173.1 | - | 8 | 1154 | |||
M15331.1 | - | 8 | 1148 | |||
M21939.1 | - | 8 | 1149 | |||
M21940.1 | - | 6 | 842 | |||
M61855.1 | - | 8 | 1175 | |||
M61857.1 | - | 8 | 1173 | |||
S46963.1 | - | 8 | 1126 |