Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007169.3 | NP_001007170.1 | 5 | 885 | ||
NM_133464.3 | NP_597721.2 | 5 | 885 | |||
XM_011518300.2 | XP_011516602.1 | 5 | 2125 | |||
XM_017014337.1 | XP_016869826.1 | 6 | 1098 | |||
XM_017014338.1 | XP_016869827.1 | 6 | 984 | |||
XM_017014339.1 | XP_016869828.1 | 5 | 885 | |||
GenBank mRNA | AB075842.1 | - | 5 | 811 | ||
AK092811.1 | - | 5 | 885 | |||
AK291267.1 | - | 5 | 877 | |||
BC065738.1 | - | 5 | 855 | |||
BC136695.1 | - | 4 | 678 | 9020685 | ||
BC136696.1 | - | 4 | 678 | 9020686 | ||
BC152427.1 | - | 5 | 811 | 8862541 | ||
BC152433.1 | - | 5 | 811 | 8862552 |