Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 9 | 1646 | ||
NM_001099773.1 | NP_001093243.1 | 9 | 1720 | |||
GenBank mRNA | AI149078.1 | - | 1 | 287 | ||
AK056794.1 | - | 9 | 1715 | |||
AK225715.1 | - | 8 | 1234 | |||
AK292300.1 | - | 9 | 1552 | |||
AK292301.1 | - | 8 | 1510 | |||
AK293211.1 | - | 10 | 1512 | |||
AK300178.1 | - | 9 | 1462 | |||
BC032329.1 | - | 9 | 1535 | 5165263 | ||
KU178011.1 | - | 9 | 1491 | |||
M14565.1 | - | 9 | 1535 | |||
M28253.1 | - | 5 | 645 |