Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM122C SIRNA Assays ›
Gene Symbol:
FAM122C
Gene Name:
family with sequence similarity 122C
Gene Aliases:
-
Chromosome Location:
Chr.X: 134796413 - 134855137 on Build GRCh38
UniGene ID:
Hs.269127
Species:
Human
siRNA ID
HSS175890
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM122C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170779.1
NP_001164250.1
2
537
NM_001170780.1
NP_001164251.1
5
444
NM_001170782.1
NP_001164253.1
3
357
NM_001170783.1
NP_001164254.1
2
537
NM_001170784.1
NP_001164255.1
3
357
NM_138819.3
NP_620174.1
2
537
XM_005262382.1
XP_005262439.1
3
315
XM_005262383.2
XP_005262440.1
3
315
XM_005262384.4
XP_005262441.1
3
315
XM_005262386.3
XP_005262443.1
3
315
XM_005262387.3
XP_005262444.1
3
315
XM_006724734.3
XP_006724797.1
3
403
XM_006724735.1
XP_006724798.1
3
315
XM_006724736.2
XP_006724799.1
3
315
XM_011531295.1
XP_011529597.1
3
315
XM_011531296.2
XP_011529598.1
3
315
XM_011531297.2
XP_011529599.1
3
315
XM_011531298.2
XP_011529600.1
3
315
XM_011531305.2
XP_011529607.1
3
315
XM_011531306.2
XP_011529608.1
3
315
XM_017029308.1
XP_016884797.1
3
315
XM_017029311.1
XP_016884800.1
3
393
GenBank mRNA
AK295485.1
-
3
295
AK307219.1
-
5
444
AK310079.1
-
2
493
BC017868.1
-
2
536
4699951
BC063491.1
-
2
477
5229041
BC065225.1
-
3
262
5724414
BG532414.1
-
2
534
DB458889.1
-
3
358
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline