Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144974.3 | NP_659411.2 | 5 | 528 | ||
XM_017020397.1 | XP_016875886.1 | 5 | 646 | |||
XM_017020398.1 | XP_016875887.1 | 5 | 646 | |||
XM_017020399.1 | XP_016875888.1 | 5 | 646 | |||
GenBank mRNA | AK056408.1 | - | 5 | 529 | ||
BC137289.1 | - | 4 | 448 | 9021286 | ||
BC137290.1 | - | 4 | 448 | 9021287 | ||
BC144570.1 | - | 4 | 448 | 9053098 | ||
BC144571.1 | - | 4 | 448 | 9053099 | ||
DA739121.1 | - | 5 | 529 |