Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141969.1 | NP_001135441.1 | 8 | 2413 | ||
NM_001141970.1 | NP_001135442.1 | 8 | 2344 | |||
NM_001254717.1 | NP_001241646.1 | 7 | 2154 | |||
NM_001350.4 | NP_001341.1 | 8 | 2410 | |||
XM_005248860.3 | XP_005248917.1 | 6 | 1260 | |||
GenBank mRNA | AA533586.1 | - | 1 | 212 | ||
AB209493.1 | - | 2 | 3232 | |||
AF006041.1 | - | 3 | 731 | |||
AF015956.2 | - | 7 | 2260 | |||
AF039136.1 | - | 8 | 2353 | |||
AF050179.1 | - | 8 | 2321 | |||
AK223404.1 | - | 8 | 2411 | |||
AK292187.1 | - | 8 | 2411 | |||
AK303767.1 | - | 8 | 2287 | |||
AK303854.1 | - | 7 | 2111 | |||
AK313034.1 | - | 8 | 2353 | |||
BC000220.1 | - | 1411 | ||||
BC109073.1 | - | 8 | 2296 | 40034214 | ||
BC109074.1 | - | 8 | 2296 | 40034219 | ||
BM997178.1 | - | 1 | 216 | |||
CR457085.1 | - | 7 | 2206 | |||
HQ436528.1 | - | 8 | 2054 | |||
HQ436529.1 | - | 7 | 2036 |