Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153813.2 | NP_722520.2 | 5 | 521 | ||
XM_011522912.2 | XP_011521214.1 | 5 | 4370 | |||
XM_011522914.2 | XP_011521216.1 | 5 | 605 | |||
XM_011522917.2 | XP_011521219.1 | 3 | 431 | |||
XM_017022982.1 | XP_016878471.1 | 3 | 310 | |||
GenBank mRNA | AF488691.1 | - | 5 | 810 |