Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DBI SIRNA Assays ›
Gene Symbol:
DBI
Gene Name:
diazepam binding inhibitor, acyl-CoA binding protein
Gene Aliases:
ACBD1, ACBP, CCK-RP, EP
Chromosome Location:
Chr.2: 119366924 - 119372551 on Build GRCh38
UniGene ID:
Hs.78888
Species:
Human
siRNA ID
HSS175958
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DBI
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079862.2
NP_001073331.1
3, 4
314
NM_001079863.1
NP_001073332.1
3, 4
434
NM_001178017.1
NP_001171488.1
3, 4
453
NM_001178041.2
NP_001171512.1
4, 5
440
NM_001178042.2
NP_001171513.1
4, 5
395
NM_001178043.2
NP_001171514.1
3, 4
344
NM_001282633.1
NP_001269562.1
4, 5
430
NM_001282634.1
NP_001269563.1
4, 5
400
NM_001282635.1
NP_001269564.1
4, 5
430
NM_001282636.1
NP_001269565.1
3, 4
344
NM_020548.7
NP_065438.1
4, 5
425
NR_104221.1
3, 4
593
GenBank mRNA
AM000001.1
-
3, 4
431
BC006466.1
-
3, 4
229
BC062996.1
-
4, 5
394
5168729
BM768076.1
-
3, 4
261
BP294452.1
-
3, 4
256
BP318721.1
-
2, 3
118
BU599304.1
-
3, 4
317
CR456956.1
-
2, 3
186
FM213122.1
-
3, 4
291
FM213123.1
-
4, 5
377
FM213124.1
-
4, 5
372
FM213125.1
-
4, 5
347
FM213126.1
-
4, 5
342
FM213127.1
-
3, 4
453
FM213128.1
-
4, 5
387
FM213129.1
-
3, 4
344
FM213130.1
-
3, 4
593
FM213131.1
-
4, 5
430
FM213132.1
-
4, 5
322
M14200.1
-
3, 4
256
M15887.1
-
3, 4
243
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
protein palmitoylation
phosphatidylcholine acyl-chain remodeling
lipid binding
benzodiazepine receptor binding
long-chain fatty acyl-CoA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline