Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005215.3 | NP_005206.2 | 6 | 1661 | ||
XM_011525843.1 | XP_011524145.1 | 6 | 1661 | |||
XM_011525844.2 | XP_011524146.1 | 3 | 549 | |||
XM_017025568.1 | XP_016881057.1 | 6 | 1661 | |||
XM_017025569.1 | XP_016881058.1 | 6 | 1661 | |||
XM_017025570.1 | XP_016881059.1 | 2 | 557 | |||
GenBank mRNA | AK302643.1 | - | 6 | 966 | ||
AK310385.1 | - | 3 | 275 | |||
BC036524.1 | - | 5 | 847 | |||
M32292.1 | - | 6 | 1051 | |||
X76132.1 | - | 6 | 1045 |