Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199085.1 | NP_001186014.1 | 12 | 2401 | ||
NM_001199089.1 | NP_001186018.1 | 12 | 2114 | |||
NM_001199091.1 | NP_001186020.1 | 12 | 2242 | |||
NM_001199092.1 | NP_001186021.1 | 9 | 1086 | |||
NM_173533.3 | NP_775804.2 | 12 | 2401 | |||
XM_005244934.1 | XP_005244991.1 | 12 | 2186 | |||
XM_005244935.4 | XP_005244992.1 | 12 | 2091 | |||
XM_017000473.1 | XP_016855962.1 | 12 | 2107 | |||
GenBank mRNA | AK092142.1 | - | 9 | 1099 | ||
AK126889.1 | - | 12 | 2242 | |||
AK127370.1 | - | 12 | 2133 | |||
BC043368.1 | - | 3 | 251 | |||
BC065707.1 | - | 9 | 1210 | |||
BC089387.1 | - | 10 | 1303 | |||
BC130532.1 | - | 12 | 2078 | 40146563 | ||
BC144058.1 | - | 12 | 2133 | 9052578 |