Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000555.3 | NP_000546.2 | 2 | 545 | ||
NM_001195553.1 | NP_001182482.1 | 2 | 247 | |||
NM_178151.2 | NP_835364.1 | 2 | 374 | |||
NM_178152.2 | NP_835365.1 | 2 | 247 | |||
NM_178153.2 | NP_835366.1 | 2 | 247 | |||
XM_011530878.2 | XP_011529180.1 | 2 | 219 | |||
XM_011530879.2 | XP_011529181.1 | 2 | 219 | |||
XM_011530880.2 | XP_011529182.1 | 2 | 219 | |||
XM_017029312.1 | XP_016884801.1 | 2 | 312 | |||
GenBank mRNA | AF034634.1 | - | 1 | 130 | ||
AF040254.1 | - | 2 | 172 | |||
AF040255.1 | - | 2 | 172 | |||
AJ003112.1 | - | 2 | 545 | |||
AK290455.1 | - | 2 | 194 | |||
AK297297.1 | - | 2 | 348 | |||
AK299822.1 | - | 2 | 196 | |||
AU120758.1 | - | 2 | 193 | |||
BC027925.1 | - | 2 | 189 | 5240179 | ||
DA497601.1 | - | 2 | 247 | |||
DC375659.1 | - | 2 | 375 |