Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317335.1 | NP_001304264.1 | 4 | 635 | ||
NM_001928.3 | NP_001919.2 | 4 | 614 | |||
GenBank mRNA | AJ313463.1 | - | 4 | 583 | ||
AK300963.1 | - | 4 | 609 | |||
BC034529.1 | - | 3 | 530 | |||
BC040146.1 | - | 3 | 516 | |||
BC051001.1 | - | 4 | 554 | |||
BC057807.1 | - | 4 | 613 | 5269709 | ||
CR536575.1 | - | 3 | 508 | |||
DQ884409.1 | - | 4 | 604 | |||
HY281047.1 | - | 2 | 395 | |||
M84526.1 | - | 4 | 562 |