Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176814.4 | NP_789784.2 | 5 | 1862 | ||
XM_005250181.3 | XP_005250238.1 | 5 | 1936 | |||
XM_006715867.3 | XP_006715930.1 | 5 | 1941 | |||
XM_006715868.3 | XP_006715931.1 | 5 | 1936 | |||
XM_006715869.3 | XP_006715932.1 | 4 | 1845 | |||
XM_011515859.2 | XP_011514161.1 | 5 | 1657 | |||
GenBank mRNA | AF218032.1 | - | 5 | 1833 | ||
AK297946.1 | - | 3 | 1050 | |||
AK309674.1 | - | 5 | 1856 | |||
BC037348.1 | - | 1 | 836 | |||
BC047641.1 | - | 5 | 1880 | |||
BC110623.1 | - | 5 | 1407 | 40036036 | ||
BC110624.1 | - | 5 | 1402 | 40036038 |