Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194294.2 | NP_919270.2 | 10 | 1101 | ||
GenBank mRNA | AK092502.1 | - | 3 | 449 | ||
AK128691.1 | - | 10 | 2730 | |||
EF052681.1 | - | 9 | 820 |