Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042413.1 | NP_001035878.1 | 9 | 3060 | ||
NM_152629.3 | NP_689842.3 | 8 | 2076 | |||
XM_005251386.4 | XP_005251443.1 | 8 | 2619 | |||
XM_005251387.4 | XP_005251444.1 | 8 | 2897 | |||
XM_005251388.4 | XP_005251445.1 | 7 | 2411 | |||
XM_011517763.2 | XP_011516065.1 | 9 | 2739 | |||
XM_011517764.2 | XP_011516066.1 | 9 | 2569 | |||
XM_011517765.2 | XP_011516067.1 | 9 | 3106 | |||
XM_011517766.2 | XP_011516068.1 | 8 | 3148 | |||
XM_011517767.2 | XP_011516069.1 | 8 | 2542 | |||
XM_017014361.1 | XP_016869850.1 | 8 | 2847 | |||
GenBank mRNA | AB209404.1 | - | 6 | 669 | ||
AK096318.1 | - | 6 | 703 | |||
BC033899.2 | - | 8 | 2076 | 4828458 | ||
DQ438877.1 | - | 2941 | ||||
DQ438878.1 | - | 6 | 1004 | |||
DQ438879.1 | - | 6 | 887 | |||
DQ438884.1 | - | 1 | 781 | |||
KU178885.1 | - | 7 | 1882 | |||
KU178886.1 | - | 7 | 1882 | |||
KU178887.1 | - | 4 | 331 |