Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042413.1 | NP_001035878.1 | 4 | 1506 | ||
NM_152629.3 | NP_689842.3 | 3 | 522 | |||
XM_005251386.4 | XP_005251443.1 | 3 | 1065 | |||
XM_005251387.4 | XP_005251444.1 | 3 | 1343 | |||
XM_005251388.4 | XP_005251445.1 | 2 | 857 | |||
XM_005251389.4 | XP_005251446.1 | 4 | 1555 | |||
XM_006716731.3 | XP_006716794.1 | 4 | 1555 | |||
XM_011517763.2 | XP_011516065.1 | 4 | 1185 | |||
XM_011517764.2 | XP_011516066.1 | 4 | 1015 | |||
XM_011517765.2 | XP_011516067.1 | 4 | 1552 | |||
XM_011517766.2 | XP_011516068.1 | 3 | 1594 | |||
XM_011517767.2 | XP_011516069.1 | 3 | 988 | |||
XM_011517769.2 | XP_011516071.1 | 4 | 1555 | |||
XM_017014361.1 | XP_016869850.1 | 3 | 1293 | |||
GenBank mRNA | BC033899.2 | - | 3 | 522 | 4828458 | |
KU178885.1 | - | 2 | 328 | |||
KU178886.1 | - | 2 | 328 |