Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130902.2 | NP_570972.2 | 3 | 363 | ||
XM_005248056.4 | XP_005248113.1 | 2 | 294 | |||
XM_011513630.2 | XP_011511932.1 | 6 | 728 | |||
XM_011513631.2 | XP_011511933.1 | 6 | 752 | |||
XM_011513632.2 | XP_011511934.1 | 5 | 652 | |||
XM_011513633.2 | XP_011511935.1 | 4 | 513 | |||
XM_011513634.2 | XP_011511936.1 | 5 | 609 | |||
XM_011513635.2 | XP_011511937.1 | 5 | 589 | |||
XM_011513636.1 | XP_011511938.1 | 2 | 252 | |||
XM_011513637.2 | XP_011511939.1 | 4 | 533 | |||
XM_011513638.2 | XP_011511940.1 | 4 | 470 | |||
GenBank mRNA | AF125109.1 | - | 3 | 311 | ||
BC035923.1 | - | 3 | 360 | 5541089 | ||
BC107855.1 | - | 4 | 434 | 5171051 |