Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134870.1 | NP_001128342.1 | 2, 3 | 1836 | ||
NM_133471.3 | NP_597728.1 | 1, 2 | 3482 | |||
GenBank mRNA | AB073600.1 | - | 2, 3 | 1727 | ||
AB075829.1 | - | 1, 2 | 2900 | |||
AK097089.1 | - | 1, 2 | 1635 | |||
AK124880.1 | - | 1, 2 | 3482 | |||
AK126182.1 | - | 803 | ||||
AL832318.1 | - | 1, 2 | 763 | |||
AL832742.1 | - | 1, 2 | 2233 | |||
BC006176.1 | - | 1, 2 | 290 | |||
BC066644.1 | - | 1, 2 | 1794 | 5575643 | ||
BX647622.1 | - | 2, 3 | 1736 | |||
DQ988947.1 | - | 1, 2 | 277 |