Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134870.1 | NP_001128342.1 | 2 | 412 | ||
NM_133471.3 | NP_597728.1 | 1 | 2058 | |||
GenBank mRNA | AB073600.1 | - | 2 | 303 | ||
AB075829.1 | - | 1 | 1476 | |||
AK097089.1 | - | 1 | 211 | |||
AK124880.1 | - | 1 | 2058 | |||
AK125639.1 | - | 2 | 1139 | |||
AK126182.1 | - | 927 | ||||
AL832742.1 | - | 1 | 810 | |||
BC066644.1 | - | 1 | 370 | 5575643 | ||
BX647622.1 | - | 2 | 312 | |||
DA788428.1 | - | 2 | 412 |