Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018050.3 | NP_001018060.1 | 5 | 798 | ||
NM_001018052.3 | NP_001018062.1 | 4 | 711 | |||
NM_001282884.1 | NP_001269813.1 | 6 | 526 | |||
NM_001282885.1 | NP_001269814.1 | 6 | 485 | |||
NM_138338.4 | NP_612211.1 | 6 | 492 | |||
XM_011529993.1 | XP_011528295.1 | 6 | 927 | |||
XM_017028641.1 | XP_016884130.1 | 7 | 1365 | |||
GenBank mRNA | AB051452.1 | - | 5 | 653 | ||
AK026524.1 | - | 6 | 472 | |||
AY092087.1 | - | 6 | 470 | |||
BC017248.2 | - | 4 | 598 | 4892259 | ||
BC088367.1 | - | 5 | 661 | 30385483 | ||
BU173504.1 | - | 5 | 440 | |||
BX331353.2 | - | 5 | 819 | |||
BX367405.2 | - | 4 | 632 | |||
CR456459.1 | - | 6 | 526 | |||
DA449644.1 | - | 7 | 507 | |||
DA504810.1 | - | 6 | 474 |