Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014762.3 | NP_055577.1 | 5, 6 | 1002 | ||
GenBank mRNA | AB073391.1 | - | 6, 7 | 877 | ||
AF261758.1 | - | 5, 6 | 972 | |||
AK302774.1 | - | 5, 6 | 833 | |||
AK304437.1 | - | 5, 6 | 719 | |||
AK316427.1 | - | 5, 6 | 703 | |||
BC004375.1 | - | 5, 6 | 908 | 3638639 | ||
BC011669.2 | - | 5, 6 | 935 | 4130396 | ||
D13643.3 | - | 5, 6 | 985 |