Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000903.2 | NP_000894.1 | 6 | 988 | ||
NM_001025433.1 | NP_001020604.1 | 5 | 886 | |||
NM_001025434.1 | NP_001020605.1 | 5 | 874 | |||
NM_001286137.1 | NP_001273066.1 | 4 | 810 | |||
GenBank mRNA | AK223050.1 | - | 6 | 900 | ||
AK297125.1 | - | 4 | 702 | |||
AK297979.1 | - | 5 | 804 | |||
AK298896.1 | - | 6 | 736 | |||
AK307533.1 | - | 6 | 850 | |||
AK309513.1 | - | 7 | 802 | |||
AK312368.1 | - | 6 | 914 | |||
AK316246.1 | - | 5 | 709 | |||
BC007659.2 | - | 6 | 914 | 3349257 | ||
CD014005.1 | - | 5 | 808 | |||
J03934.1 | - | 6 | 847 |