Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006729.4 | NP_006720.1 | 13 | 1814 | ||
NM_007309.3 | NP_009293.1 | 13 | 1814 | |||
GenBank mRNA | AK002029.1 | - | 1 | 86 | ||
AK291272.1 | - | 13 | 1696 | |||
BC117414.1 | - | 14 | 1481 | 40125965 | ||
BC143838.1 | - | 14 | 1481 | 9052353 | ||
Y15908.1 | - | 13 | 1768 | |||
Y15909.1 | - | 13 | 1768 |