Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052838.4 | NP_443070.5 | 6 | 528 | ||
XM_017022991.1 | XP_016878480.1 | 6 | 508 | |||
GenBank mRNA | AF085235.1 | - | 5 | 362 | ||
AK301045.1 | - | 5 | 360 | |||
AK303390.1 | - | 6 | 491 | |||
AL833004.1 | - | 2 | 107 | |||
AY034176.1 | - | 5 | 362 | |||
BC012161.1 | - | 4 | 350 | 4576220 | ||
BQ072902.1 | - | 6 | 541 |