Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000792.6 | NP_000783.2 | 1 | 135 | ||
NM_001039715.2 | NP_001034804.1 | 1 | 135 | |||
NM_001039716.2 | NP_001034805.1 | 1 | 135 | |||
NM_001324316.1 | NP_001311245.1 | 1 | 135 | |||
NM_213593.4 | NP_998758.1 | 1 | 135 | |||
NR_136692.1 | 1 | 135 | ||||
NR_136693.1 | 1 | 135 | ||||
GenBank mRNA | AK290780.1 | - | 1 | 135 | ||
AY560374.1 | - | 1 | 111 | |||
AY560375.1 | - | 1 | 111 | |||
AY560376.1 | - | 1 | 111 | |||
AY560377.1 | - | 1 | 111 | |||
AY560378.1 | - | 1 | 111 | |||
AY560379.1 | - | 1 | 111 | |||
AY560381.1 | - | 1 | 111 | |||
AY560382.1 | - | 1 | 111 | |||
AY560383.1 | - | 1 | 111 | |||
BC017955.1 | - | 1 | 134 | 4607072 | ||
BC107170.2 | - | 1 | 134 | 40036824 | ||
BC107171.2 | - | 1 | 134 | 40036826 | ||
CD014030.1 | - | 1 | 117 | |||
FJ002243.1 | - | 1 | 133 | |||
FJ002244.1 | - | 1 | 133 | |||
FJ002245.1 | - | 1 | 133 | |||
FJ002247.1 | - | 1 | 133 | |||
FJ002248.1 | - | 1 | 133 |