Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005221.5 | NP_005212.1 | 1 | 480 | ||
GenBank mRNA | AK023493.1 | - | 1 | 480 | ||
AK297614.1 | - | 1 | 600 | |||
BC006226.2 | - | 1 | 466 | 3941691 | ||
BT006903.1 | - | 1 | 272 | |||
DB280023.1 | - | 1 | 480 |