Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMBT1 SIRNA Assays ›
Gene Symbol:
DMBT1
Gene Name:
deleted in malignant brain tumors 1
Gene Aliases:
GP340, SAG, muclin
Chromosome Location:
Chr.10: 122560665 - 122643736 on Build GRCh38
UniGene ID:
Hs.279611
Species:
Human
siRNA ID
HSS176203
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMBT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320644.1
NP_001307573.1
40
4985
NM_004406.2
NP_004397.2
27
3101
NM_007329.2
NP_015568.2
40
4985
NM_017579.2
NP_060049.2
39
4955
XM_006717660.3
XP_006717723.1
42
5312
XM_006717665.3
XP_006717728.1
41
5264
XM_011539388.2
XP_011537690.1
43
5360
XM_011539389.2
XP_011537691.1
43
5360
XM_011539390.2
XP_011537692.1
42
5330
XM_011539391.2
XP_011537693.1
42
5330
XM_011539392.2
XP_011537694.1
42
5330
XM_011539393.2
XP_011537695.1
42
5330
XM_011539394.2
XP_011537696.1
42
5330
XM_011539395.2
XP_011537697.1
42
5330
XM_011539396.2
XP_011537698.1
42
5312
XM_011539398.2
XP_011537700.1
41
5297
XM_011539399.2
XP_011537701.1
41
5264
XM_011539400.2
XP_011537702.1
42
5036
XM_011539401.2
XP_011537703.1
40
4973
XM_011539402.2
XP_011537704.1
40
4973
XM_011539403.2
XP_011537705.1
40
4973
XM_011539405.2
XP_011537707.1
40
4974
XM_011539407.2
XP_011537709.1
40
4967
XM_011539408.2
XP_011537710.1
40
4967
XM_011539409.2
XP_011537711.1
40
4967
XM_011539411.2
XP_011537713.1
39
4943
XM_011539413.2
XP_011537715.1
37
4579
XM_011539414.2
XP_011537716.1
27
3080
XM_017015798.1
XP_016871287.1
28
3622
GenBank mRNA
AF159456.1
-
40
4985
AJ000342.1
-
27
3101
AJ243212.1
-
40
4985
AJ243224.1
-
39
4955
AJ297935.1
-
40
4966
BC153299.1
-
24
2719
40147087
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
pattern recognition receptor signaling pathway
receptor-mediated endocytosis
multicellular organism development
protein transport
epithelial cell differentiation
induction of bacterial agglutination
cellular protein metabolic process
innate immune response
defense response to virus
scavenger receptor activity
protein binding
signaling pattern recognition receptor activity
zymogen binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-5xwc9:80/10.239.165.207:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0