Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081560.2 | NP_001075029.1 | 7 | 1002 | ||
NM_001081562.2 | NP_001075031.1 | 7 | 1002 | |||
NM_001081563.2 | NP_001075032.1 | 6 | 1371 | |||
NM_001288764.1 | NP_001275693.1 | 8 | 1061 | |||
NM_001288765.1 | NP_001275694.1 | 5 | 642 | |||
NM_001288766.1 | NP_001275695.1 | 7 | 1002 | |||
NM_004409.4 | NP_004400.4 | 7 | 1002 | |||
GenBank mRNA | AB209363.1 | - | 7 | 1845 | ||
AK297299.1 | - | 5 | 593 | |||
AK302874.1 | - | 7 | 935 | |||
BC026328.1 | - | 8 | 1056 | 4792104 | ||
BC062553.1 | - | 7 | 923 | 6480902 | ||
KJ534826.1 | - | 6 | 827 | |||
KJ534827.1 | - | 6 | 827 | |||
KJ534828.1 | - | 6 | 827 | |||
L19268.1 | - | 7 | 924 | |||
M87312.1 | - | 6 | 1372 | |||
M94203.1 | - | 7 | 674 | |||
S72883.1 | - | 7 | 833 |