Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009932.2 | NP_001009932.1 | 7 | 1014 | ||
NM_001009933.2 | NP_001009933.1 | 6 | 756 | |||
NM_001009934.2 | NP_001009934.1 | 5 | 671 | |||
NM_001303620.1 | NP_001290549.1 | 5 | 637 | |||
NM_006730.3 | NP_006721.1 | 5 | 1191 | |||
XM_005277829.4 | XP_005277886.1 | 7 | 1148 | |||
XM_011531122.2 | XP_011529424.1 | 6 | 1697 | |||
XM_017029332.1 | XP_016884821.1 | 7 | 2220 | |||
GenBank mRNA | BC001561.1 | - | 5 | 555 | 3456429 | |
BC028092.1 | - | 5 | 1191 | 5219882 | ||
BT019990.1 | - | 4 | 383 | |||
L40823.1 | - | 6 | 716 | |||
U06846.1 | - | 5 | 574 | |||
X90392.1 | - | 6 | 1177 |
Gene Symbol: RPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927570.1 | - | LIT3571 AY927570 |
4 | 564 |