Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207055.1 | NP_001193984.1 | 15 | 1936 | ||
NM_001207056.1 | NP_001193985.1 | 14 | 1834 | |||
NM_006892.3 | NP_008823.1 | 17 | 2122 | |||
NM_175848.1 | NP_787044.1 | 16 | 2062 | |||
NM_175849.1 | NP_787045.1 | 16 | 2062 | |||
NM_175850.2 | NP_787046.1 | 16 | 2041 | |||
XM_011528653.2 | XP_011526955.1 | 16 | 1914 | |||
XM_011528654.2 | XP_011526956.1 | 15 | 1789 | |||
GenBank mRNA | AB208880.1 | - | 16 | 2286 | ||
AF156487.1 | - | 16 | 1855 | |||
AF156488.1 | - | 17 | 1915 | |||
AF176228.1 | - | 16 | 2041 | |||
AF331857.1 | - | 17 | 2122 | |||
AK001191.1 | - | 10 | 994 | |||
AK295176.1 | - | 14 | 1536 | |||
AK299821.1 | - | 14 | 1577 | |||
AK299915.1 | - | 15 | 1713 | |||
DQ321787.1 | - | 16 | 2156 |