Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004947.4 | NP_004938.1 | 35 | 3614 | ||
XM_005264914.3 | XP_005264971.1 | 35 | 3886 | |||
XM_005264915.3 | XP_005264972.1 | 35 | 3886 | |||
XM_005264916.3 | XP_005264973.1 | 34 | 3637 | |||
XM_005264917.3 | XP_005264974.1 | 35 | 3844 | |||
XM_005264918.3 | XP_005264975.1 | 34 | 3516 | |||
XM_006713008.3 | XP_006713071.1 | 35 | 3886 | |||
XM_006713009.3 | XP_006713072.1 | 35 | 3886 | |||
XM_006713010.3 | XP_006713073.1 | 35 | 3649 | |||
XM_011533441.2 | XP_011531743.1 | 24 | 4495 | |||
XM_011533443.2 | XP_011531745.1 | 11 | 2176 | |||
XM_011533444.2 | XP_011531746.1 | 11 | 2100 | |||
XM_011533445.2 | XP_011531747.1 | 11 | 2121 | |||
XM_017005825.1 | XP_016861314.1 | 35 | 3665 | |||
XM_017005826.1 | XP_016861315.1 | 35 | 3655 | |||
XM_017005827.1 | XP_016861316.1 | 26 | 2806 | |||
XM_017005828.1 | XP_016861317.1 | 10 | 1531 | |||
GenBank mRNA | AB002297.1 | - | 30 | 3222 | ||
AY145303.1 | - | 35 | 3591 | |||
AY254099.1 | - | 35 | 3614 |