Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001387.2 | NP_001378.1 | 1, 2 | 152 | ||
GenBank mRNA | AI570709.1 | - | 2, 3 | 304 | ||
BC039006.1 | - | 1, 2 | 118 | 5725550 | ||
D78014.1 | - | 1, 2 | 152 | |||
Y07818.1 | - | 1, 2 | 173 |