Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171184.1 | NP_001164655.1 | 10 | 1281 | ||
NM_001939.2 | NP_001930.2 | 12 | 1564 | |||
XM_017029334.1 | XP_016884823.1 | 11 | 1643 | |||
GenBank mRNA | AB209305.1 | - | 3 | 161 | ||
AK289825.1 | - | 12 | 1452 | |||
AK295843.1 | - | 10 | 1150 | |||
BC111695.1 | - | 12 | 1401 | 40036070 | ||
U43519.1 | - | 12 | 1513 |