Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285389.2 | NP_001272318.1 | 4 | 448 | ||
NM_001285391.2 | NP_001272320.2 | 4 | 771 | |||
NM_001285392.2 | NP_001272321.1 | 4 | 565 | |||
NM_001285393.2 | NP_001272322.1 | 4 | 502 | |||
NM_004414.6 | NP_004405.3 | 4 | 774 | |||
NM_203417.2 | NP_981962.1 | 4 | 560 | |||
NM_203418.2 | NP_981963.1 | 4 | 739 | |||
XM_017028283.1 | XP_016883772.1 | 5 | 1100 | |||
GenBank mRNA | AF400429.1 | - | 4 | 432 | ||
AK055845.1 | - | 3 | 971 | |||
AK092184.1 | - | 4 | 695 | |||
AK131569.1 | - | 4 | 692 | |||
AK299444.1 | - | 4 | 489 | |||
AL556803.3 | - | 4 | 774 | |||
AY325903.1 | - | 4 | 766 | |||
BC002864.2 | - | 4 | 534 | 3944959 | ||
BT007363.1 | - | 4 | 478 | |||
BX419260.2 | - | 4 | 692 | |||
CR456878.1 | - | 3 | 400 | |||
CR542065.1 | - | 4 | 478 | |||
DA586027.1 | - | 4 | 502 | |||
U28833.2 | - | 4 | 448 | |||
U85265.2 | - | 4 | 559 | |||
U85266.2 | - | 4 | 502 | |||
U85267.2 | - | 4 | 621 |