Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005609.1 | NP_001005609.1 | 1 | 607 | ||
NM_001005610.3 | NP_001005610.2 | 1 | 607 | |||
NM_001005612.2 | NP_001005612.2 | 1 | 607 | |||
NM_001005613.3 | NP_001005613.1 | 1 | 607 | |||
NM_001399.4 | NP_001390.1 | 1 | 607 | |||
XM_006724630.2 | XP_006724693.1 | 1 | 588 | |||
XM_011530885.2 | XP_011529187.1 | 1 | 588 | |||
XM_017029336.1 | XP_016884825.1 | 1 | 588 | |||
XM_017029337.1 | XP_016884826.1 | 1 | 588 | |||
GenBank mRNA | AF040628.1 | - | 1 | 607 | ||
AF060999.1 | - | 1 | 606 | |||
AF061189.1 | - | 1 | 607 | |||
AF061190.1 | - | 1 | 607 | |||
AF061191.1 | - | 1 | 607 | |||
AF061192.1 | - | 1 | 607 | |||
AF061193.1 | - | 1 | 607 | |||
AF061194.1 | - | 1 | 607 | |||
BC126143.1 | - | 1 | 557 | 8991859 | ||
BC144049.1 | - | 1 | 557 | 9052569 | ||
BC144051.1 | - | 1 | 557 | 9052571 | ||
CA433299.1 | - | 2 | 248 | |||
U59228.1 | - | 1 | 607 |