Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004427.3 | NP_004418.2 | 5 | 856 | ||
NM_198040.2 | NP_932157.1 | 12 | 2163 | |||
XM_005270570.1 | XP_005270627.1 | 5 | 792 | |||
XM_017000513.1 | XP_016856002.1 | 13 | 2662 | |||
XM_017000514.1 | XP_016856003.1 | 13 | 2274 | |||
XM_017000515.1 | XP_016856004.1 | 13 | 2367 | |||
XM_017000516.1 | XP_016856005.1 | 13 | 2596 | |||
XM_017000517.1 | XP_016856006.1 | 13 | 2208 | |||
XM_017000518.1 | XP_016856007.1 | 13 | 2301 | |||
XM_017000519.1 | XP_016856008.1 | 13 | 2671 | |||
XM_017000520.1 | XP_016856009.1 | 12 | 2121 | |||
XM_017000521.1 | XP_016856010.1 | 12 | 2540 | |||
GenBank mRNA | AJ419231.1 | - | 12 | 2163 | ||
AK056429.1 | - | 6 | 1093 | |||
AK128821.1 | - | 5 | 674 | |||
AK292905.1 | - | 13 | 2601 | |||
AK294426.1 | - | 7 | 1441 | |||
BC018602.1 | - | 5 | 862 | |||
BC028396.1 | - | 5 | 827 | |||
BC029269.1 | - | 5 | 698 | |||
BC068573.1 | - | 5 | 750 | |||
BC092492.1 | - | 5 | 750 | |||
BC110863.1 | - | 5 | 749 | |||
BC130630.1 | - | 11 | 2079 | 40146661 | ||
BC144121.1 | - | 11 | 2079 | 9052641 | ||
U89278.1 | - | 5 | 847 |