Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
UniGene ID:
Hs.535192
Species:
Human
siRNA ID
HSS176311
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001402.5
NP_001393.1
6
958
XM_011535514.2
XP_011533816.1
6
1593
GenBank mRNA
AF174496.1
-
5
788
AF328730.1
-
1
666
AF397403.1
-
4
703
AJ420488.1
-
5
918
AK024888.1
-
1
956
AK026650.1
-
1
958
AK055058.1
-
6
958
AK058051.1
-
6
958
AK058163.1
-
5
776
AK098510.1
-
6
958
AK098518.1
-
6
818
AK098587.1
-
3
385
AK098623.1
-
6
958
AK127384.1
-
6
958
AK222515.1
-
1
958
AK222519.1
-
1
607
AK222523.1
-
1
958
AK222551.1
-
1
958
AK222982.1
-
1
958
AK223030.1
-
1
955
AK223042.1
-
1
813
AK223046.1
-
1
961
AK289374.1
-
6
958
AK289442.1
-
6
896
AK290691.1
-
6
958
AK292639.1
-
6
1274
AK297878.1
-
7
756
AK300922.1
-
6
819
AK304214.1
-
7
918
AK315827.1
-
6
949
AY043301.1
-
5
1305
BC008587.1
-
1
944
3447654
BC009733.1
-
1
908
3877516
BC009875.2
-
1
948
3948601
BC010735.1
-
1
937
3898364
BC012891.1
-
1
936
3858483
BC014224.2
-
1
947
4335756
BC014377.1
-
1
370
BC014892.1
-
1
254
BC018150.1
-
1
944
3851939
BC018641.2
-
1
944
3660140
BC018850.2
-
1
947
BC019050.2
-
1
910
BC020477.2
-
1
607
BC021686.1
-
1
963
3930863
BC022412.1
-
1
259
BC028674.1
-
1
962
4107346
BC029337.1
-
1
932
BC029343.1
-
1
930
BC029997.1
-
1
945
BC035877.1
-
1
959
BC038339.1
-
1
944
3461588
BC057391.1
-
1
913
4339606
BC063511.1
-
1
381
BC065761.1
-
1
263
BC066893.1
-
1
958
4818863
BC071619.1
-
2
874
4448926
BC071727.1
-
1
913
4514479
BC071741.1
-
1
938
3885539
BC071841.1
-
1
933
4587981
BC072385.1
-
1
945
5585092
BC082268.1
-
1
937
6729595
BC094687.1
-
2
850
30366186
BC111051.1
-
1
948
5585079
EF219140.1
-
6
1118
EF362804.1
-
6
1188
M27364.1
-
2
196
M29548.1
-
3
490
X03558.1
-
6
948
X16869.1
-
5
910
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline