Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024852.3 | NP_079128.2 | 15 | 2325 | ||
NM_177422.2 | NP_803171.1 | 13 | 2067 | |||
XM_005270575.3 | XP_005270632.1 | 16 | 7779 | |||
XM_005270576.3 | XP_005270633.1 | 10 | 2563 | |||
XM_011540879.2 | XP_011539181.1 | 15 | 3207 | |||
XM_011540882.2 | XP_011539184.1 | 12 | 1422 | |||
XM_017000523.1 | XP_016856012.1 | 16 | 7795 | |||
XM_017000524.1 | XP_016856013.1 | 11 | 1593 | |||
XM_017000525.1 | XP_016856014.1 | 12 | 1427 | |||
XM_017000526.1 | XP_016856015.1 | 11 | 1386 | |||
XM_017000527.1 | XP_016856016.1 | 11 | 1565 | |||
XM_017000528.1 | XP_016856017.1 | 12 | 1514 | |||
GenBank mRNA | AK022827.1 | - | 15 | 2080 | ||
AK292870.1 | - | 15 | 2291 | |||
AK302289.1 | - | 8 | 1074 | |||
AK303929.1 | - | 12 | 1609 | |||
BC025769.1 | - | 13 | 1891 |