Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178111.1 | NP_001171582.1 | 4 | 326 | ||
NM_001178112.1 | NP_001171583.1 | 4 | 293 | |||
NM_138967.3 | NP_620417.1 | 3 | 243 | |||
NR_033660.1 | 3 | 243 | ||||
XM_006720420.1 | XP_006720483.1 | 3 | 402 | |||
XM_006720421.1 | XP_006720484.1 | 4 | 284 | |||
XM_006720422.3 | XP_006720485.1 | 4 | 252 | |||
XM_017021989.1 | XP_016877478.1 | 3 | 392 | |||
GenBank mRNA | AB105192.1 | - | 3 | 214 | ||
AF495715.1 | - | 3 | 195 | |||
AK056438.1 | - | 4 | 268 | |||
AK126940.1 | - | 3 | 243 | |||
AK127480.1 | - | 4 | 263 | |||
AK301505.1 | - | 3 | 205 | |||
AL834226.1 | - | 3 | 214 | |||
BC024700.1 | - | 2 | 86 | 4941050 | ||
CR936761.1 | - | 3 | 176 | |||
DA810757.1 | - | 3 | 243 |