Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1D SIRNA Assays ›
Gene Symbol:
EEF1D
Gene Name:
eukaryotic translation elongation factor 1 delta
Gene Aliases:
EF-1D, EF1D, FP1047
Chromosome Location:
Chr.8: 143579697 - 143597675 on Build GRCh38
UniGene ID:
Hs.333388
Species:
Human
siRNA ID
HSS176328
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130053.2
NP_001123525.2
8
1888
NM_001130055.2
NP_001123527.1
7
959
NM_001130056.2
NP_001123528.1
5
711
NM_001130057.2
NP_001123529.1
6
965
NM_001195203.1
NP_001182132.1
6
726
NM_001289950.1
NP_001276879.1
6
780
NM_001317743.1
NP_001304672.1
5
708
NM_001960.4
NP_001951.2
6
783
NM_032378.4
NP_115754.3
8
2005
XM_005250820.3
XP_005250877.1
7
6880
XM_005250823.1
XP_005250880.1
7
1614
XM_006716519.3
XP_006716582.1
7
6877
XM_006716520.3
XP_006716583.1
6
6808
XM_006716522.1
XP_006716585.1
7
1868
XM_006716523.1
XP_006716586.1
8
1828
XM_006716524.1
XP_006716587.1
7
1635
XM_006716525.1
XP_006716588.1
6
803
XM_011516905.2
XP_011515207.1
8
6450
XM_011516906.2
XP_011515208.1
10
5042
XM_017013168.1
XP_016868657.1
8
5881
XM_017013169.1
XP_016868658.1
9
5537
XM_017013170.1
XP_016868659.1
8
5323
XM_017013171.1
XP_016868660.1
7
1611
XM_017013172.1
XP_016868661.1
6
1796
XM_017013173.1
XP_016868662.1
5
888
GenBank mRNA
AF370363.1
-
8
1588
AK024550.1
-
8
1742
AK093756.1
-
6
1925
AK225040.1
-
8
1743
AK225243.1
-
6
520
AK225270.1
-
6
521
AK293339.1
-
6
464
AK314311.1
-
8
1624
AL833379.1
-
2
3255
BC000678.2
-
5
1255
BC007847.2
-
8
1609
4129368
BC009907.2
-
1
505
BC012819.2
-
1
505
BC062535.1
-
6
497
BC071840.1
-
6
485
BC094806.1
-
5
422
BG482145.1
-
5
415
BQ064320.1
-
640
BQ686171.1
-
7
678
BT007242.1
-
6
1546
CN424105.1
-
6
568
Z21507.1
-
6
517
Back To Top
More Information
Panther Classification:
Molecular Function -
translation elongation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to ionizing radiation
cell-cell adhesion
DNA binding
translation elongation factor activity
signal transducer activity
protein binding
translation factor activity, RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-nvdhv:80/10.239.163.173:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0