Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030059.2 | NP_001025230.1 | 3 | 581 | ||
NM_001318166.1 | NP_001305095.1 | 3 | 581 | |||
NM_001318167.1 | NP_001305096.1 | 3 | 581 | |||
XM_005269592.2 | XP_005269649.1 | 3 | 581 | |||
XM_006717685.3 | XP_006717748.1 | 3 | 250 | |||
XM_006717686.3 | XP_006717749.1 | 3 | 581 | |||
XM_011539444.2 | XP_011537746.1 | 4 | 308 | |||
XM_011539445.2 | XP_011537747.1 | 3 | 581 | |||
XM_017015820.1 | XP_016871309.1 | 3 | 253 | |||
GenBank mRNA | AK098668.1 | - | 3 | 199 | ||
AK296273.1 | - | 3 | 288 | |||
AK310178.1 | - | 3 | 232 | |||
BC101268.2 | - | 4 | 275 | 40024099 | ||
BC132787.1 | - | 3 | 404 | 40146809 | ||
BI550027.1 | - | 3 | 313 | |||
CR739184.1 | - | 2 | 194 | |||
DB473853.2 | - | 3 | 288 |