Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304274.1 | NP_001291203.1 | 2 | 174 | ||
NM_144981.2 | NP_659418.1 | 3 | 237 | |||
XM_005252812.3 | XP_005252869.1 | 4 | 358 | |||
XM_011519942.1 | XP_011518244.1 | 4 | 295 | |||
XM_011519943.2 | XP_011518245.1 | 5 | 416 | |||
XM_011519944.2 | XP_011518246.1 | 4 | 324 | |||
XM_011519945.2 | XP_011518247.1 | 3 | 239 | |||
XM_011519946.2 | XP_011518248.1 | 4 | 284 | |||
XM_011519947.2 | XP_011518249.1 | 3 | 221 | |||
XM_017017305.1 | XP_016872794.1 | 3 | 295 | |||
XM_017017307.1 | XP_016872796.1 | 2 | 151 | |||
GenBank mRNA | AK057788.1 | - | 3 | 231 | ||
AK310496.1 | - | 2 | 154 | |||
BC023595.2 | - | 2 | 120 | 4841844 | ||
DB288287.1 | - | 4 | 296 |