Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113402.1 | NP_001106873.1 | 2 | 260 | ||
NM_001278411.1 | NP_001265340.1 | 2 | 240 | |||
NM_001278412.1 | NP_001265341.1 | 4 | 469 | |||
XM_017018963.1 | XP_016874452.1 | 4 | 309 | |||
XM_017018964.1 | XP_016874453.1 | 2 | 242 | |||
XM_017018965.1 | XP_016874454.1 | 3 | 334 | |||
GenBank mRNA | AK123033.1 | - | 2 | 107 | ||
AK302112.1 | - | 4 | 316 | |||
AL833434.1 | - | 2 | 110 | |||
BC045831.1 | - | 2 | 204 | 5265088 | ||
BI464068.1 | - | 2 | 189 | |||
DC325581.1 | - | 2 | 166 | |||
DC394802.1 | - | 2 | 260 |